Description
The Thermo Scientific HPLC system is applied for the analysis of triazine herbicides. The separation is performed on a Thermo Scientific Acclaim 120 C18 column, with MS detection.| Market: | Environmental |
| Keywords: | Acclaim 120 C18, Ametryn, Atrazine, HPLC, LCMS, MS detection, Prometon, Propazine, Simazine, Terbutryn, Triazine herbicides |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.