Find methods for your needs
Analysis of alkaloids in bitter orange extract by HPLC-UV
Description
The Thermo Scientific HPLC system is applied for the analysis of alkaloids in bitter orange extract which is an ingredient in traditional Chinese medicines. The separation is performed on a Thermo Scientific Acclaim 120 C18 column, with UV detection at 224 nm and is suitable for assaying ingredients and formulations for bitter orange and its active constituents. Chromatogram A is standards and chromatogram B is an extract of bitter orange.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, Alkaloids, Bitter orange, Hordenine, HPLC, N-Methyltyramine, Octopamine, Phenylephrine, Synephrine, Tyramine, UV detection |
| Matrix: | Bitter orange extract |
For Research Use Only. Not for use in diagnostic procedures.