Description
The Thermo Scientific HPLC system is applied for the analysis of nonylphenol isomers. The separation is performed on a Thermo Scientific Hypercarb Kappa column, with UV detection at 204 nm.| Market: | Environmental |
| Keywords: | HPLC, Hypercarb, Nonylphenol isomers, UV detection, Kappa |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.