Find methods for your needs
Rapid analysis of allantoin by HPLC-UV Be the first to rate this application
rapid analysis allantoin by hplcuv
Description
The Thermo Scientific HPLC system is applied for the analysis of allantoin. The separation is performed on a Thermo Scientific Hypercarb column in less than 5 minutes, with UV detection at 205 nm.
Market: Pharma
Keywords: Allantoin, HPLC, Hypercarb, UV detection
Matrix: N/A
Uploaded on 6/19/2015.
For Research Use Only. Not for use in diagnostic procedures.