Find methods for your needs
Rapid analysis of nine corticosteroids using a Thermo Scientific Acclaim RSLC C18 columm Be the first to rate this application
Description
The Thermo Scientific Ultimate 3000 system is applied for the rapid analysis of nine corticosteroids. The separation was performed with a Thermo Scientific Acclaim RSLC C18 column with UV detection at 254 nm
Market: Pharma
Keywords: Acclaim 120 C18, Betamethasone, Corticosteroids, Cortisone, Cortisone acetate, Dexamethasone, HPLC, Hydrocortisone, Methylprednisone, Prednisolone, Prednisone, Ttiamcinolone acetate, UV detection
Matrix: N/A
Uploaded on 2/25/2011.
For Research Use Only. Not for use in diagnostic procedures.