Description
In this work the suitability and selective performance of the Thermo Scientific TraceGOLD TG-5SilMS for the analysis of total organic carbon was demonstrated. TOC can be used as a useful measure of the broad estimation or residual pesticides and other carbonaceous material. It is especially useful when coupled to mass selective detection as it allows identification and quantification of specific contaminating materials. Note the retention times in the component table match the component number and are not the actual retention time.| Market: | Environmental |
| Keywords: | Chlorinated pesticides, GC, MS detection, Organic pollutants, Organophosphorous pesticides, Polyaromatic hydrocarbons, Total organic carbon (TOC), TraceGOLD TG-5SilMS, Triazines, US EPA 508, US EPA 608, US EPA 610, US EPA 619, US EPA 8080, US EPA 8081, US EPA 8100, US EPA 8140, US EPA 8141, US EPA 8141A |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.