Find methods for your needs
EPA 8015 analysis of aliphatic hydrocarbons in diesel and gasoline range organics in ground and surface water by GC/MS
Description
The Thermo Scientific TRACE GC Ultra is applied for the analysis of aliphatic hydrocarbons in diesel and gasoline range organics in ground and surface water by EPA method 8015. The separation was performed on a Thermo Scientific TraceGOLD TG-5SilMS GC column, with MS detection.| Market: | Environmental |
| Keywords: | Aliphatic hydrocarbons, Diesel range organics, EPA 8015, Gasoline range organics, GC-MS, GRO, Ground water, Surface water, TraceGOLD TG-5SilMS |
| Matrix: | Ground, Surface water |
For Research Use Only. Not for use in diagnostic procedures.