Find methods for your needs
Simple extraction of vanillin from a cola soft drink using SPE with analysis by GC-MS
Description
The Thermo Scientific TRACE GC Ultra is applied for the analysis of vanillin from a cola soft drink. The sample was prepared using Thermo Scientific SOLA HRP solid phase extraction (SPE) cartridges, and the separation was performed on a Thermo Scientific TraceGOLD TG-WaxMS A GC column, with flame ionization detection.| Market: | Food and Beverage |
| Keywords: | Cola, GC, Soft drink, SOLA HRP, SPE, TraceGOLD TG-WaxMS, Vanillin, MS detection |
| Matrix: | Cola |
For Research Use Only. Not for use in diagnostic procedures.