 Rate
                        Rate
                     Ask the Expert
                        Ask the Expert
                     Suggest an Application
                            Suggest an Application
                         Add to Favorites
                        Add to Favorites
                     Create
                            CreateParts List
 Tell a
                        Tell a Friend
 Print
                        PrintFind methods for your needs
Description
In this work, the performance of a 0.15 mm internal diameter (ID) GC column is compared with that of a 0.25 mm ID GC column. A 50% increase in the speed of analysis for a 14 component fatty acid methyl ester (FAME) reference standard C8–C24 mix is demonstrated using the 0.15 mm GC column. No compromise in the separation capability of the method was observed, and conversion of the conventional method was easily achieved using conventional GC instrumentation.| Market: | Food and Beverage | 
| Keywords: | FAMEs, Fatty acid methyl esters, FID detection, GC, TraceGOLD TG-WaxMS | 
| Matrix: | N/A | 
For Research Use Only. Not for use in diagnostic procedures.