Find methods for your needs
Fast analysis of organochlorine pesticides by GC-FID Be the first to rate this application
fast analysis organochlorine pesticides by gcfid
Description
In this work, the performance of a 0.15 mm internal diameter (ID) Thermo Scientific TraceGOLD TG-5SilMS GC column is compared with that of a 0.25 mm ID GC column. An increase in the speed of analysis for an organochlorine pesticide (OCP) reference standard mix using EPA Method 8081 is demonstrated with the 0.15 mm column. No compromise in the separation capability of the method was observed and conversion of the conventional method was easily achieved.
Market: Environmental
Keywords: FID detection, GC, Organochlorine pesticides, TraceGOLD TG-5SilMS
Matrix: N/A
Uploaded on 5/19/2015.
For Research Use Only. Not for use in diagnostic procedures.