Find methods for your needs
Simple and fast QuEChERS dispersive solid phase extraction for the GC-MS analysis of pesticides in cucumber
Description
The Thermo Scientific TRACE GC Ultra is applied for the analysis of pesticides residues. The pesticides were extracted from cucumber using the QuEChERS method described in EN15662. The separation of the pesticides was performed on a Thermo Scientific TraceGOLD TG-5MS GC column, with MS detection.| Market: | Food and Beverage |
| Keywords: | GC, Cucumber, MS detection, Pesticides, QuEChERS, TraceGOLD TG-5MS |
| Matrix: | Cucumber |
For Research Use Only. Not for use in diagnostic procedures.