Find methods for your needs
Demonstration of EPA Method 8141B for the high resolution separation of organophosphorus pesticides by GC-FID
Description
The TraceGOLD TG-5MS column and the deactivated, splitless quartz liner with quartz wool, when used in a TRACE 1310 GC instrument, demonstrated excellent performance for the separation and analysis of organophosphorus compounds with excellent peak shape, resolution, and reproducibility.| Market: | Environmental |
| Keywords: | FID detection, GC, Organophosphorus pesticides, TraceGOLD TG-5MS, EPA 8141B |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.