Find methods for your needs
AN140: Fast Analysis of Anions in Drinking Water by Ion Chromatography
Description
The U.S. National Primary Drinking Water Standards specify a Maximum Contaminant Level (MCL) for a number of inorganic anions to minimize potential health effects. In this Application Note, we describe the use of the IonPac AS14A anion exchange column for the routine high throughput determination of common inorganic anions in drinking water matrices. The IonPac AS14A provides greater speed and efficiency, ruggedness equivalent to the AS4A-SC column, improved separation of fluoride from the void volume (water dip), and better overall separation selectivity.| Market: | Environmental |
| Keywords: | Chloride, Carbonate, sulfate, fluoride, Phosphate, Bromide, Nitrate, IonPac AS14A, EPA method 300.0 |
| Matrix: | Drinking water |
| Author: | Valoran Hanko and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.