Find methods for your needs
Improved separation of tetra-alkylammonium salts by HPLC with suppressed conductivity detection Be the first to rate this application
improved separation tetraalkylammonium salts by hplc with suppressed conductivity detection
Description
The Thermo Scientific HPLC system is applied for the analysis of tetra-alkylammonium salts. The separation is performed on a Thermo Scientific Acclaim Surfactant column, with suppressed conductivity detection.
Market: Pharma
Keywords: Acclaim Surfactant, HPLC, Suppressed conductivity detection, Tetra-alkylammonium salts, Tetrabutylammonium chloride, Tetraheptylammonium bromide, Tetrahexylammonium bromide, Tetrapentylammonium bromide
Matrix: N/A
Uploaded on 5/4/2015.
For Research Use Only. Not for use in diagnostic procedures.