Find methods for your needs
Separation of cationic surfactants with superior peak shape Be the first to rate this application
separation cationic surfactants with superior peak shape
Description
The Thermo Scientific HPLC system is applied for the analysis of cationic surfactants. The separation is performed on a Thermo Scientific Acclaim Surfactant column, with suppressed conductivity detection.
Market: Pharma
Keywords: Acclaim Surfactant, Cationic surfactant, Cetylpyridinium chloride, Cetyltrimethylammonium chloride, HPLC, Lauryl pyridinium chloride, Lauryldimethylbenzyl ammonium chloride, Octylphenoxyethoxyethyl dimethylbenzyl ammonium, Suppressed conductivity detection
Matrix: N/A
Uploaded on 5/4/2015.
For Research Use Only. Not for use in diagnostic procedures.