Find methods for your needs
Good separation of quaternary surfactants by HPLC with suppressed conductivity detection Be the first to rate this application
good separation quaternary surfactants by hplc with suppressed conductivity detection
Description
The Thermo Scientific HPLC system is applied for the analysis of quaternary surfactants. The separation is performed on a Thermo Scientific Acclaim Surfactant column, with suppressed conductivity detection.
Market: Pharma
Keywords: Acclaim Surfactant, Dodecylpyridinium chloride, Hexadecylpyridinium chloride, Hexadecyl-trimethylammonium bromide, HPLC, Quaternary surfactants, Suppressed conductivity detection
Matrix: N/A
Uploaded on 5/4/2015.
For Research Use Only. Not for use in diagnostic procedures.