Find methods for your needs
Straightforward isocratic method for the analysis of phenytoin and related substances
Description
The Thermo Scientific HPLC system is applied for the analysis of phenytoin and related substances. The separation is performed on a Thermo Scientific Acclaim 120 C18 column, with UV detection at 220 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, Benzaldehyde, Benzil, Benzoic acid, Benzoin, Benzophenone, HPLC, Impurities, Phenytoin, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.