Description
The Thermo Scientific HPLC system is applied for the analysis of nitrosamines in EPA 8270. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage (PA) column, with UV detection at 230 nm.| Market: | Environmental |
| Keywords: | Acclaim PolarAdvantage, EPA 8270, HPLC, Nitrosamines, Nitroso-dibutylamine, Nitroso-diethylamine, Nitroso-dimethylamine, Nitroso-diphenylamine, Nitroso-dipropylamine, Nitroso-methylethylamine, Nitroso-morpholine, Nitroso-piperidine, Nitroso-pyrrolidine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.