Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of hydrocortisone in topical cream. The separation is performed using a Thermo Scientific Acclaim 120 C18 column, with UV detection at 245 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, HPLC, Hydrocortisone, Methyl paraben, Propyl paraben, Topical cream, UV detection |
| Matrix: | Topical cream |
For Research Use Only. Not for use in diagnostic procedures.