Find methods for your needs
Targeted analysis of 6 pesticides in milk utilising a QuEChERS sample preparation approach and GC-MS detection
Description
The Thermo Scientific Trace 13xx series GC with ISQ is applied for the detection of pesticides in milk. Sample analysis was performed utilising a QuEChERS extraction/cleanup workflow followed by separation on a Thermo Scientific TraceGOLD TG-5SilMS 30m x 0.25mm, 0.25µm column with MS detection.| Market: | Food and Beverage |
| Keywords: | Cypermethrin, GC/MS, MS detection, Pesticides, QuEChERS, Milk, Diazinon, Methyl parathion, Malathion, Chlorpyrifos, Ethion |
| Matrix: | Milk |
| Author: | Richard Law |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.