Find methods for your needs
A simple isocratic method for the determination of bimatoprost and its impurities using revered-phase HPLC-UV
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of bimatoprost and its impurities. The separation was performed on a Thermo Scientific Accucore Phenyl-Hexyl HPLC column using a simple isocratic reversed-phase method.| Market: | Pharma |
| Keywords: | Accucore Phenyl-Hexyl, Bimatoprost, HPLC, Isomers, UV detection, Impurities |
| Matrix: | N/A |
| Author: | Anirudha Rakhe |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.