Find methods for your needs
Improved separation of alkyl quaternary amines by HPLC with conductivity detection
Description
The Thermo Scientific system is applied for the analysis of alkyl quaternary amines. The separation is performed on a Thermo Scientific Acclaim Surfactant Plus HPLC column, with suppressed conductivity detection.| Market: | Other |
| Keywords: | Acclaim Surfactant Plus, Alkyl quaternary amines, Decyl-trimethylammonium, Dodecyl-trimethylammonium, Hexadecyl-trimethylammonium, Surfactant, TetrabutylammoniumTetrapentylammonium, Tetradecyl-trimethylammonium, Tetraheptylammonium, Tetrahexylammonium |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.