Find methods for your needs
Fast determination of α-tocopherol and α-tocopheryl acetate using a Thermo Scientific Acclaim HILIC-10 column Be the first to rate this application
fast determination αtocopherol αtocopheryl acetate using a thermo scientific acclaim hilic10 column
Description
The Thermo Scientific HPLC system is applied for the determination of α-tocopherol and α-tocopheryl acetate. The separation is performed on a Thermo Scientific Acclaim HILIC-10 column, with UV detection at 280 nm.
Market: Food and Beverage
Keywords: Acclaim HILIC-10, HPLC, Tocopherol, Tocopheryl, UV detection
Matrix: N/A
Uploaded on 3/4/2015.
For Research Use Only. Not for use in diagnostic procedures.