Find methods for your needs
Rapid analysis of OSHA-55 cyanocrylate adhesives using a Thermo Scientific Acclaim RSLC 120 C18 column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of OSHA-55 cyanocrylate adhesives. The separation is performed on a Thermo Scientific Acclaim 120 C18 RSLC column, with UV detection at 220 nm.| Market: | Material Science |
| Keywords: | Acclaim 120 C18, Cyanocrylate adhesives, Cynergy, Ethoxyethyl cyanoacrylate, Ethyl cyanoacrylate, HPLC, Methyl cyanoacrylate, OSHA-55, Permabond, UV detection |
| Matrix: | Adhesives |
For Research Use Only. Not for use in diagnostic procedures.