Find methods for your needs
Fast analysis of organic acids in white wine on a Thermo Scientific Acclaim Organic Acid (OA) column
Description
The Thermo Scientific HPLC system is applied for the analysis of organic acids in white wine. The separation is performed on a Thermo Scientific Acclaim Organic Acid (OA) column, with UV detection at 210 nm. The samples were prepared using Thermo Scientific Dionex OnGuard II P SPE cartridges.| Market: | Food and Beverage |
| Keywords: | Acclaim Organic Acid, Citric acid, HPLC, Lactic acid, Malic acid, Organic acids, Succinic acid, Tartaric acid, UV detection, White wine |
| Matrix: | White wine |
For Research Use Only. Not for use in diagnostic procedures.