Find methods for your needs
Rapid analysis of yellow dye in breakfast cereal using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of yellow dye in breakfast cereal. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 427, 508 and 625 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, Cereal, HPLC, UV detection, Yellow dye |
| Matrix: | Cereal |
For Research Use Only. Not for use in diagnostic procedures.