Find methods for your needs
Fast analysis of sudan dyes using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of sudan dyes. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 495 nm.| Market: | Material Science |
| Keywords: | Acclaim 120 C18, HPLC, Sudan dyes, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.