Find methods for your needs
Improved separation of phthalate esters in EPA method 606 using a Thermo Scientific PolarAdvantage (PA) column
Description
The Thermo Scientific HPLC system is applied for the determination of phthalate esters in EPA method 606. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage (PA) HPLC column, with UV detection at 254 nm.| Market: | Environmental |
| Keywords: | Acclaim PolarAdvantage (PA), EPA 606, HPLC, Phthalate esters, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.