Find methods for your needs
Improved selectivity for sigma peptide standards on a Thermo Scientific Acclaim 300 C18 column with TFA
Description
The Thermo Scientific HPLC system is applied for the analysis of sigma peptide standard with TFA as the mobile phase modifier. The separation is performed on a Thermo Scientific Acclaim 300 C18 HPLC column, with UV detection at 214 nm. Trifluoroacetic acid (TFA) is used as a mobile phase modifier in peptide analysis because it promotes efficient peak shape, in part by suppressing interactions with silanols on the column.| Market: | BioPharma |
| Keywords: | Acclaim 300 C18, HPLC, Peptides, Sigma, TFA, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.