Find methods for your needs
Improved analysis of resveratrol and related substances in dietary supplements using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the determination of resveratrol and related substances in dietary supplements. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 310 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, Dietary supplements, HPLC, Polydatin, Pterostilbene, Resveratrol, UV detection |
| Matrix: | Dietary supplements |
For Research Use Only. Not for use in diagnostic procedures.