Find methods for your needs
Fast analysis of hydroxybenzoic acids using a Thermo Scientific Acclaim Organic Acid (OA) HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of hydroxybenzoic acids. The separation is performed on a Thermo Scientific Acclaim Organic Acid (OA)HPLC column, with UV detection at 230 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim Organic Acid, Benzoic acid, HPLC, Hydroxybenzoic acids, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.