Find methods for your needs
Fast analysis of reverse-transcriptase inhibitors using a Thermo Scientific Acclaim PolarAdvantage (PA) column
Description
The Thermo Scientific HPLC system is applied for the determination of reverse-transcriptase inhibitors. The separation is performed on a Thermo Scientific Acclaim PolarAdvantage (PA) HPLC column, with UV detection at 265 nm.| Market: | Pharma |
| Keywords: | Acclaim PolarAdvantage (PA), AIDS, HIV, HPLC, Lamivudine, Nevirapine, Niacin, Reverse-transcriptase inhibitors, Stavudine, Thynine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.