Find methods for your needs
Fast and easy determination of multiple mycotoxins in grain using QuEChERS and a Thermo Scientific Accucore aQ HPLC column
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the determination of multiple mycotoxins in grain-based food. The samples are prepared using Thermo Scientific HyperSep Dispersive SPE (QuEChERS), and the separation performed using a Thermo Scientific Accucore aQ HPLC column, with MS/MS detection.| Market: | Food and Beverage |
| Keywords: | Alflatoxins, Dispersive SPE, Grain, HPLC, LC-MS/MS, Mycotoxins, Ochratoxin, QuEChERS |
| Matrix: | Grain |
For Research Use Only. Not for use in diagnostic procedures.