Find methods for your needs
Fast analysis of phenols using a Thermo Scientific Acclaim 120 C18 column
Description
The Thermo Scientific HPLC system is applied for the analysis of phenols. The separation is performed using a Thermo Scientific Acclaim 120 C18 column, with UV detection at 285 nm.| Market: | Environmental |
| Keywords: | Acclaim 120 C18, EPA 604, HPLC, Phenols, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.