Find methods for your needs
Fast analysis of EPA 604 phenols using a Thermo Scientific Acclaim RSLC PolarAdvantage (PA) column Be the first to rate this application
fast analysis epa 604 phenols using a thermo scientific acclaim rslc polaradvantage pa column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of EPA 604 phenols. The separation is performed using a Thermo Scientific Acclaim RSLC PolarAdvantage (PA) column, with UV detection at 280 nm.
Market: Environmental
Keywords: Acclaim PolarAdvantage (PA), EPA 604, HPLC, Phenols, UV detection
Matrix: N/A
Uploaded on 2/18/2015.
For Research Use Only. Not for use in diagnostic procedures.