Find methods for your needs
Fast analysis of capsaicin in chilies using a Thermo Scientific Acclaim RSLC 120 C18 column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the analysis of capsaicin in chilies. The separation is performed using a Thermo Scientific Acclaim RSLC 120 C18 column, with UV detection at 280 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, Capsaicin, Chili, Dihydrocapsaicin, HPLC, UV detection |
| Matrix: | Chili |
For Research Use Only. Not for use in diagnostic procedures.