Find methods for your needs
AN286: Determination of Trace Copper, Nickel, and Zinc in Boiling Water Reactors Using Nonsuppressed Conductivity Detection
Description
This work describes the determination of trace concentrations of copper, nickel, and zinc in a boiling water reactor (BWR) matrix (i.e., deionized water) by cation-exchange chromatography using a 2 mm Thermo Scientific Dionex IonPac SCS 1 column and nonsuppressed conductivity detection. A Thermo Scientific Dionex Ultralow Pressure Trace Cation Concentrator (TCC-ULP1) column is used to concentrate 8 mL of sample, which enables detection limits of <0.2 μg/L for copper, nickel, and zinc.Market: | Water Analysis |
Keywords: | copper, IonPac SCS 1, nickel, Nuclear Power, Oxalic acid, TCC-ULP1, zinc |
Matrix: | Water |
Author: | Brian De Borba and Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.