Find methods for your needs
IonCount solution reference test for Acclaim Trinity P2 gradient elutions Be the first to rate this application
Description
IonCount includes a complete quaternary UltiMate 3000 Standard system and a chromatography starter kit. This kit includes two different analytical columns providing alternative selectivity, all required buffer salts for the mobile phases as well as reference standards. The standards in combination with pre-defined eWorkflows allow verifying the isocratic and gradient separation methods once they are established on your UltiMate 3000 system. The standards also allow continuously monitoring system and column performance ensuring high quality data.
Market: Pharma
Keywords: Charged aerosol detection, Corona Veo, counter ion, Counterion, ion, IonCount
Matrix: N/A
Uploaded on 2/5/2015.
For Research Use Only. Not for use in diagnostic procedures.