Find methods for your needs
Application Note 65: Analysis of Inositol Phosphates Be the first to rate this application
application note 65 analysis inositol phosphates
Description
This report describes the use of high-performance anion-exchange chromatography with chemically suppressed conductivity detection to determine inositol phosphate isomers.
Market: BioPharma
Keywords: brain, IC, inositol triphosphate, INP1,4,5, OmniPac PAX-500, suppressed conductivity
Matrix: Tissue
Uploaded on 1/22/2015.
For Research Use Only. Not for use in diagnostic procedures.