Find methods for your needs
Fluconazole tablets assay according to the Chinese Pharmacopoeia using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of fluconazole tablets accordign to the Chinese Pharmacopoeia. The separation is performed on a Thermo Scientific Acclaim 120 C18 HPLC column, with UV detection at 261 nm.Market: | Pharma |
Keywords: | Acclaim 120 C18, Chinese pharmacopeia, Fluconazole, HPLC, Tablet, UV detection |
Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.