Find methods for your needs
Simple isocratic assay analysis of Levetiracetam using reversed-phase HPLC-UV
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied for the determination of levetiracetam. The assay was performed on a Thermo Scientific Accucore XL C18 HPLC column in less than 20 minutes, with UV detection at 200 nm.| Market: | Pharma |
| Keywords: | Accucore XL C18, HPLC, Levetiracetam, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.