Find methods for your needs
Accurate analysis of paracetamol and impurities in tablet formulation using a reversed-phase HPLC-UV method
Description
The Thermo Scientific Dionex Ultimate 3000 system is applied for the accurate assay and impurity analysis of paracetamol tablets. The separation is performed on a Thermo Scientific Syncronis C18 HPLC column in less than 20 minutes, with UV detection at 245 and 275 nm.| Market: | Pharma |
| Keywords: | 4-Aminophenol, 4-Chloroacetate, 4-Nitrophenol, HPLC, Impurities, Paracetamol, Tablet, UV detection |
| Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.