Find methods for your needs
Analysis of Fatty Acid Methyl Esters (FAMEs) C8-C24 by GC-FID Using a Metal Column
Description
The Thermo Scientific TraceGOLD Metal GC columns provide a robust alternative to the traditional fused silica columns, assuring longer lifetime suitable for high throughput. In this application, their performances have been proved with the separation of FAMEs in the range C8-C24, showing equivalent selectivity of a fused silica column.| Market: | Food and Beverage |
| Keywords: | Biodiesel, FAME, Fat, Fatty acid methyl esters, TG-WaxMT |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.