Find methods for your needs
高效液相色谱-紫外检测法测定山金连流感软胶囊中的绿原酸、黄芩苷和牛蒡苷 Be the first to rate this application
高效液相色谱紫外检测法测定山金连流感软胶囊中的绿原酸、黄芩苷和牛蒡苷
Description
对于含有不同紫外吸收特征组分的样品检测时,在没有DAD检测器时可以通过VWD的波长切换功能达到用一个条件完成测试的目的。测试条件2在较短时间内完成了对三个成分的测定,进样量小节约溶剂,在用户样品量较多时是很好的选择。在日常的方法开发中,如果有条件可以尽量使用小颗粒填料的色谱柱,以获得更高的灵敏度、更好的峰形和环境友好。
Market: Pharma
Keywords: 高效液相色谱;紫外检测;绿原酸;黄芩苷;牛蒡苷;山金连流感软胶囊
Matrix: 山金连流感软胶囊
Uploaded on 12/10/2014.
For Research Use Only. Not for use in diagnostic procedures.