Find methods for your needs
Rapid determination of Docetaxel in formulation using a Thermo Scientific Hypersil GOLD HPLC column Be the first to rate this application
Description
The method here demonstrates the use of the Thermo Scientific Hypersil GOLD HPLC column for the analysis of docetaxel in a formulation. The separation was performed on a Thermo Scientific Dionex Ultimate 3000 RSLC system with UV detection at 232 nm.
Market: Pharma
Keywords: Docetaxel, HPLC, Hypersil GOLD, Taxane, UV detection
Matrix: Formulation
Uploaded on 12/3/2014.
For Research Use Only. Not for use in diagnostic procedures.