Find methods for your needs
加速溶剂萃取-顺序萃取法萃取陈皮中的橙皮苷 Be the first to rate this application
加速溶剂萃取顺序萃取法萃取陈皮中的橙皮苷
Description
采用ASE顺序提取法提取陈皮中的橙皮苷,相比较2010版药典推荐方法-索氏提取,能够达到了除脂和萃取有效成分的目的,并简化了操作步骤及节省了大量的操作时间。ASE提取中药中的活性成分时,既能达到药典推荐萃取方法等同的萃取率,在提高萃取条件时,ASE还能达到更高的萃取率。同时具有操作简单、快速、高效等特点,值得在中药领域推广使用。
Market: Pharma
Keywords: 加速溶剂萃取;顺序提取;中药;陈皮;橙皮苷
Matrix: 石油醚、甲醇
Uploaded on 12/3/2014.
For Research Use Only. Not for use in diagnostic procedures.