Find methods for your needs
Simple isocratic separation of acetamiprid and methylparaben using HPLC-UV
Description
The Thermo Scientific Dionex UltiMate 3000 system is applied for the analysis of an insecticide and an anti-fungal agent. The Thermo Scientific Acclaim 120 C18 5 µm column separated acetamiprid and methylparaben in 12 minutes with UV detection at 250 nm.| Market: | Environmental |
| Keywords: | Acclaim 120 C18, Acetamiprid, antifungal, HPLC, Insecticide, Methylparaben, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.