Find methods for your needs
Rapid separation of microcystins using a Thermo Scientific Acclaim RSLC C18 column
Description
The Thermo Scientific UlitiMate 3000 system is applied for the rapid analysis of microcystins. The separation of three variants of microcystins was performed using the Thermo Scientific Acclaim RSLC C18 2.2 µm with UV detection at 238 nm.| Market: | Water Analysis |
| Keywords: | Acclaim 120 C18, HPLC, Microcystins, Toxin, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.