Find methods for your needs
Separation of Ketoconazole in anti-dandruff shampoo using a Thermo Scientific Acclaim 120 C18 HPLC column
Description
The analysis of anti-dandruff shampoo was performed using the Thermo Scientific Acclaim 120 C18 3 μm column with UV detection at 223 nm.| Market: | Pharma |
| Keywords: | Anti-Dandruff Shampoo, antifungal, HPLC, Ketoconazole, UV detection |
| Matrix: | Shampoo |
For Research Use Only. Not for use in diagnostic procedures.