Find methods for your needs
An improved separation of isoflavones in red clover using a Thermo Scientific Acclaim 120 C18 HPLC column Be the first to rate this application
an improved separation isoflavones red clover using a thermo scientific acclaim 120 c18 hplc column
Description
The Thermo Scientific Dionex Accelerated Solvent Extractor system (ASE) system was used to extract the red clover samples with reduced time and solvent use. Thermo Scientific Acclaim C18 5 μm column was used to separate the isoflavones extracted from red clover in a run time under 25 minutes.
Market: Environmental
Keywords: Accelerated Solvent Extractor, Biochanin A, Formononotin, Genistin, HPLC, Isoflavones, Ononin, Red Clover, Sissotrin, UV detection
Matrix: Red clover
Uploaded on 11/7/2014.
For Research Use Only. Not for use in diagnostic procedures.